(1-Decyl)triphenylphosphonium bromide
Supplier: Apollo Scientific
|
Warning
|
OR18367-100G
OR18367-1KG
APOSOR18367-100GEA
261
EUR
APOSOR18367-100G
APOSOR18367-1KG
(1-Decyl)triphenylphosphonium bromide
(1-Decyl)triphenylphosphonium bromide
Formula:
C₂₈H₃₆BrP Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051854 CAS Number: 32339-43-8 EINECS: 250-996-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...