Iminodiacetonitrile, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
OR18385-2.5KG
OR18385-500G
APOSOR18385-2.5KGEA
405
EUR
APOSOR18385-2.5KG
APOSOR18385-500G
Iminodiacetonitrile, Technical Grade
Iminodiacetonitrile
Formula:
C₄H₅N₃ MW: 95,11 g/mol Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001887 CAS Number: 628-87-5 EINECS: 211-058-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...