2-Methoxy-6-methylbenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR18565-10G OR18565-5G
APOSOR18565-10GEA 74.2 EUR
APOSOR18565-10G APOSOR18565-5G
2-Methoxy-6-methylbenzoic acid 98%
2-Methoxy-6-methylbenzoic acid
Formula: C₉H₁₀O₃
MW: 166,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463871
CAS Number: 6161-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR