5-Amino-2-nitropyridine 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Nitropyridin-3-amine
OR18602-5G
OR18602-1G
OR18602-250MG
APOSOR18602-5GEA
205
EUR
APOSOR18602-5G
APOSOR18602-1G
APOSOR18602-250MG
5-Amino-2-nitropyridine 95%
5-Amino-2-nitropyridine
Formula:
C₅H₅N₃O₂ MW: 139.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01646117 CAS Number: 14916-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...