Carbazole 98%

Supplier: Apollo Scientific
Warning

OR1866-25G OR1866-100G
APOSOR1866-25GEA 105 EUR
APOSOR1866-25G APOSOR1866-100G
Carbazole 98%
Carbazole
Formula: C₁₂H₉N
MW: 167,21 g/mol
Boiling Pt: 355 °C
Melting Pt: 240…246 °C
Density: 1,1 g/cm³ (25 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR