4-Methyl-1H-1,2,3-benzotriazole 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Methyl-1H-benzo[d][1,2,3]triazole
OR18853-1G
OR18853-5G
APOSOR18853-1GEA
335
EUR
APOSOR18853-1G
APOSOR18853-5G
4-Methyl-1H-1,2,3-benzotriazole 95%
4-Methyl-1H-1,2,3-benzotriazole
Formula:
C₇H₇N₃ MW: 133,15 g/mol Melting Pt: 141…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035800 CAS Number: 29878-31-7 EINECS: 249-921-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...