3-Bromoquinoline
Supplier: Apollo Scientific
|
Warning
|
OR1946-100G
OR1946-25G
APOSOR1946-100GEA
52.4
EUR
APOSOR1946-100G
APOSOR1946-25G
3-Bromoquinoline
3-Bromoquinoline
Formula:
C₉H₆BrN MW: 208,06 g/mol Boiling Pt: 274…276 °C Melting Pt: 13…15 °C Density: 1,533 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006767 CAS Number: 5332-24-1 EINECS: 226-237-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...