1-Propanesulphonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Propane-1-sulphonyl chloride
OR19492-500G
APOSOR19492-500GEA
94.4
EUR
APOSOR19492-500G
1-Propanesulphonyl chloride 97%
1-Propanesulphonyl chloride
Formula:
C₃H₇ClO₂S MW: 142,61 g/mol Boiling Pt: 78…79 °C/15mmHg Density: 1,267 g/cm³ (25 °C) Flash Pt: 97 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007462 CAS Number: 10147-36-1 EINECS: 233-414-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...