HA14-1 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl-2-amino-6-bromo-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate, rel-(R)-Ethyl 2-amino-6-bromo-4-((R)-1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate(relative)
OR200014-1G
OR200014-250MG
APOSOR200014-1GEA
376
EUR
APOSOR200014-1G
APOSOR200014-250MG
HA14-1 95%
HA14-1
Formula:
C₁₇H₁₇BrN₂O₅ MW: 409,24 g/mol Melting Pt: 105…108 °C Storage Temperature: Freezer |
MDL Number:
MFCD00218213 CAS Number: 65673-63-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...