2-Chloro-1H-benzimidazole-6-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-1H-benzo[d]imidazole-5-carbonitrile
OR200030-250MG
OR200030-100MG
OR200030-500MG
APOSOR200030-250MGEA
401
EUR
APOSOR200030-250MG
APOSOR200030-100MG
APOSOR200030-500MG
2-Chloro-1H-benzimidazole-6-carbonitrile
2-Chloro-1H-benzimidazole-6-carbonitrile
Formula:
C₈H₄ClN₃ MW: 177,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08457981 CAS Number: 401567-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...