5-Chloro-6-methoxypyridine-3-carboxylic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloro-6-methoxynicotinic acid

OR200033-1G
APOSOR200033-1GEA 347 EUR
APOSOR200033-1G
5-Chloro-6-methoxypyridine-3-carboxylic acid 96%
5-Chloro-6-methoxypyridine-3-carboxylic acid
Formula: C₇H₆ClNO₃
MW: 187,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972405
CAS Number: 884494-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR