Pyrimidine-4-carbonyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR200047-1G
OR200047-250MG
OR200047-1MG
OR200047-5MG
OR200047-10MG
OR200047-500MG
APOSOR200047-1GEA
643
EUR
APOSOR200047-1G
APOSOR200047-250MG
APOSOR200047-1MG
APOSOR200047-5MG
APOSOR200047-10MG
APOSOR200047-500MG
Pyrimidine-4-carbonyl chloride
Pyrimidine-4-carbonyl chloride
Formula:
C₅H₃ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD12827989 CAS Number: 184951-32-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...