Thioridazine hydrochloride 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
10-[2-(1-Methylpiperidin-2-yl)ethyl]-2-(methylsulphanyl)-10H-phenothiazine hydrochloride
OR20007-25G
OR20007-5G
APOSOR20007-25GEA
377
EUR
APOSOR20007-25G
APOSOR20007-5G
Thioridazine hydrochloride 98%
Thioridazine hydrochloride
Formula:
C₂₁H₂₇ClN₂S₂ MW: 407.031 g/mol Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012655 CAS Number: 130-61-0 EINECS: 204-992-8 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...