(4-Bromophenyl)acetaldehyde 95%

Supplier: Apollo Scientific
Warning

OR200079-1G OR200079-5G
APOSOR200079-1GEA 45.1 EUR
APOSOR200079-1G APOSOR200079-5G
(4-Bromophenyl)acetaldehyde 95%
(4-Bromophenyl)acetaldehyde
Formula: C₈H₇BrO
Boiling Pt: 113…114 °C/7mmHg
Melting Pt: 96…97 °C
Storage Temperature: Ambient
MDL Number: MFCD02261728
CAS Number: 27200-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR