(2-Chloropyridin-4-yl)methylamine 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(2-Chloropyridin-4-yl)methanamine, 4-Aminomethyl-2-chloropyridine
OR20014-1G
OR20014-5G
APOSOR20014-1GEA
0
EUR
APOSOR20014-1G
APOSOR20014-5G
(2-Chloropyridin-4-yl)methylamine 95%
(2-Chloropyridin-4-yl)methylamine
Formula:
C₆H₇ClN₂ MW: 142,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06212875 CAS Number: 144900-57-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...