Methyl 6-bromohexanoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromo-hexanoic acid methyl ester

OR2002-100G OR2002-25G OR2002-500G
APOSOR2002-100GEA 58 EUR
APOSOR2002-100G APOSOR2002-25G APOSOR2002-500G
Methyl 6-bromohexanoate 98%
Methyl 6-bromohexanoate
Formula: C₇H₁₃BrO₂
MW: 209.084 g/mol
Boiling Pt: 70…74 °C/1mmHg
Density: 1.316 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00187528
CAS Number: 14273-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR