2-(Hydroxymethyl)indole 97+%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(Indole-2-yl)methanol, 2-Indolmethanol, Indole-2-methanol, Indole-2-carbinol, 1H-Indole-2-methanol, (1H-Indol-2-yl)methanol, 2-Indolcarbinol
OR20024-25G
APOSOR20024-25GEA
97.3
EUR
APOSOR20024-25G
2-(Hydroxymethyl)indole 97+%
2-(Hydroxymethyl)indole
Formula:
C₉H₉NO MW: 147,18 g/mol Melting Pt: 72…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00030195 CAS Number: 24621-70-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...