7-Chlorothieno[3,2-b]pyridine 98%

Supplier: Apollo Scientific
Warning

OR20039-5G OR20039-1G
APOSOR20039-5GEA 25.8 EUR
APOSOR20039-5G APOSOR20039-1G
7-Chlorothieno[3,2-b]pyridine 98%
7-Chlorothieno[3,2-b]pyridine
Formula: C₇H₄ClNS
Storage Temperature: Refrigerator
MDL Number: MFCD08448161
CAS Number: 69627-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR