2'-Bromoacetophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: o-Bromoacetophenone

OR2031-100G OR2031-25G OR2031-500G
APOSOR2031-100GEA 116 EUR
APOSOR2031-100G APOSOR2031-25G APOSOR2031-500G
2'-Bromoacetophenone 98%
2'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199,05 g/mol
Density: 1,476 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0
EINECS: 218-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR