1H-Benzimidazole-5-carbaldehyde 97+%

Supplier: Apollo Scientific
Warning

OR2063-1G OR2063-250MG
APOSOR2063-1GEA 161 EUR
APOSOR2063-1G APOSOR2063-250MG
1H-Benzimidazole-5-carbaldehyde 97+%
1H-Benzimidazole-5-carbaldehyde
Formula: C₈H₆N₂O
MW: 146,15 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD08741423
CAS Number: 58442-17-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR