Nicotinic acid hydrazide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide
OR21092-100G
OR21092-25G
APOSOR21092-100GEA
60.9
EUR
APOSOR21092-100G
APOSOR21092-25G
Nicotinic acid hydrazide
Nicotinic acid hydrazide
Formula:
C₆H₇N₃O MW: 137,14 g/mol Melting Pt: 159…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006383 CAS Number: 553-53-7 EINECS: 209-041-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...