Isoniazid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

OR21093-100G OR21093-25G
APOSOR21093-100GEA 24.8 EUR
APOSOR21093-100G APOSOR21093-25G
Isoniazid
Isoniazid
Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 169…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006426
CAS Number: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR