Ethyl-2-chloro-5-nitrobenzoate
Supplier: Apollo Scientific
OR21154-1G
OR21154-5G
APOSOR21154-1GEA
65.5
EUR
APOSOR21154-1G
APOSOR21154-5G
Ethyl-2-chloro-5-nitrobenzoate
Ethyl-2-chloro-5-nitrobenzoate
Formula:
C₉H₈ClNO₄ MW: 229,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00115803 CAS Number: 16588-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...