1-(4-Nitrophenyl)piperidine

Supplier: Apollo Scientific

OR21172-1G OR21172-5G OR21172-10G
APOSOR21172-1GEA 42.2 EUR
APOSOR21172-1G APOSOR21172-5G APOSOR21172-10G
1-(4-Nitrophenyl)piperidine
1-(4-Nitrophenyl)piperidine
Formula: C₁₁H₁₄N₂O₂
MW: 206,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023662
CAS Number: 6574-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR