3-Bromoaniline 98%
Supplier: Apollo Scientific
|
Danger
|
OR2121-100G
OR2121-25G
APOSOR2121-100GEA
64
EUR
APOSOR2121-100G
APOSOR2121-25G
3-Bromoaniline 98%
3-Bromoaniline
Formula:
C₆H₆BrN MW: 172,02 g/mol Boiling Pt: 74…75 °C/1mmHg Melting Pt: 15…18 °C Density: 1,58 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007757 CAS Number: 591-19-5 EINECS: 209-704-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...