Methyl-2-amino-4-chlorobenzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-amino-4-chlorobenzoate
OR21323-100G
APOSOR21323-100GEA
106
EUR
APOSOR21323-100G
Methyl-2-amino-4-chlorobenzoate
Methyl 2-amino-4-chlorobenzoate
Formula:
C₈H₈ClNO₂ MW: 185,61 g/mol Boiling Pt: 106 °C/8mmHg Melting Pt: 65…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017568 CAS Number: 5900-58-3 EINECS: 227-593-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...