α-Bromo-3,4-dichlorotoluene 97%

Supplier: Apollo Scientific
Danger

Synonyms: 3,4-Dichlorobenzyl bromide

OR21377-25G OR21377-5G
APOSOR21377-25GEA 78.3 EUR
APOSOR21377-25G APOSOR21377-5G
α-Bromo-3,4-dichlorotoluene 97%
α-Bromo-3,4-dichlorotoluene
Formula: C₇H₅BrCl₂
MW: 239,93 g/mol
Boiling Pt: 115…120 °C/1mmHg
Density: 1,65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041363
CAS Number: 18880-04-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR