2',5'-Dimethylacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2,5-Dimethylphenyl)ethanone, 1-(2,5-Dimethylphenyl)ethan-1-one
OR21390-25G
OR21390-5G
APOSOR21390-25GEA
77
EUR
APOSOR21390-25G
APOSOR21390-5G
2',5'-Dimethylacetophenone 98%
2',5'-Dimethylacetophenone
Formula:
C₁₀H₁₂O MW: 148,2 g/mol Boiling Pt: 230…231 °C Flash Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014988 CAS Number: 2142-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...