3-Chlorobenzamide 98%
Supplier: Apollo Scientific
|
Warning
|
OR21585-100G
OR21585-25G
APOSOR21585-100GEA
310
EUR
APOSOR21585-100G
APOSOR21585-25G
3-Chlorobenzamide 98%
3-Chlorobenzamide
Formula:
C₇H₆ClNO Melting Pt: 133…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014799 CAS Number: 618-48-4 EINECS: 210-554-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...