2,4-Dibromotoluene 98%

Supplier: Apollo Scientific
Warning

OR2172-25G OR2172-5G
APOSOR2172-25GEA 39.3 EUR
APOSOR2172-25G APOSOR2172-5G
2,4-Dibromotoluene 98%
2,4-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249,93 g/mol
Boiling Pt: 243 °C
Melting Pt: –10 °C
Density: 1,85 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052985
CAS Number: 31543-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR