Methyl (3-methoxyphenyl)acetate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3-Methoxyphenylacetate
OR21784-1G
OR21784-25G
OR21784-5G
APOSOR21784-1GEA
14.7
EUR
APOSOR21784-1G
APOSOR21784-25G
APOSOR21784-5G
Methyl (3-methoxyphenyl)acetate 98%
Methyl (3-methoxyphenyl)acetate
Formula:
C₁₀H₁₂O₃ MW: 180,23 g/mol Boiling Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017205 CAS Number: 18927-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...