2-(3-Methoxyphenyl)acetohydrazide
Supplier: Apollo Scientific
|
Warning
|
OR21788-1G
OR21788-5G
APOSOR21788-1GEA
43.5
EUR
APOSOR21788-1G
APOSOR21788-5G
2-(3-Methoxyphenyl)acetohydrazide
2-(3-Methoxyphenyl)acetohydrazide
Formula:
C₉H₁₂N₂O₂ MW: 180,21 g/mol Melting Pt: 89…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085106 CAS Number: 34624-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...