Methyl-4-amino-2-methoxybenzoate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 4-amino-2-methoxybenzenecarboxylate, 4-Amino-2-methoxybenzoic acid methyl ester, Methyl 4-amino-2-methoxybenzoate
OR21849-100G
OR21849-25G
OR21849-500G
APOSOR21849-100GEA
55.1
EUR
APOSOR21849-100G
APOSOR21849-25G
APOSOR21849-500G
Methyl-4-amino-2-methoxybenzoate 98%
Methyl 4-amino-2-methoxybenzoate
Formula:
C₉H₁₁NO₃ Melting Pt: 155…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017202 CAS Number: 27492-84-8 EINECS: 248-494-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...