5-Methylnicotinic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Methyl-3-pyridinecarboxylic acid, 5-Methylpyridine-3-carboxylic acid

OR21864-1G OR21864-25G OR21864-5G
APOSOR21864-1GEA 14.7 EUR
APOSOR21864-1G APOSOR21864-25G APOSOR21864-5G
5-Methylnicotinic acid 97%
5-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137,14 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00829036
CAS Number: 3222-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR