3-Amino-2-nitropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Nitro-3-pyridylamine, 2-Nitropyridin-3-amine, 2-Nitropyridin-3-ylamine
OR21871-100G
APOSOR21871-100GEA
192
EUR
APOSOR21871-100G
3-Amino-2-nitropyridine 98%
3-Amino-2-nitropyridine
Formula:
C₅H₅N₃O₂ MW: 139,11 g/mol Melting Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044102 CAS Number: 13269-19-7 EINECS: 236-260-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...