4-Butylbenzoyl chloride 98%
Supplier: Apollo Scientific
|
Danger
|
OR21934-25G
APOSOR21934-25GEA
0
EUR
APOSOR21934-25G
4-Butylbenzoyl chloride 98%
4-Butylbenzoyl chloride
Formula:
C₁₁H₁₃ClO MW: 196,68 g/mol Boiling Pt: 145…147 °C/11mmHg Density: 1,051 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000699 CAS Number: 28788-62-7 EINECS: 249-224-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...