2-Allyl-3-hydroxybenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR21984-100MG
OR21984-1G
APOSOR21984-100MGEA
29
EUR
APOSOR21984-100MG
APOSOR21984-1G
2-Allyl-3-hydroxybenzaldehyde
2-Allyl-3-hydroxybenzaldehyde
Formula:
C₁₀H₁₀O₂ Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100646 CAS Number: 79950-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...