2,6-Dibromoaniline 98%

Supplier: Apollo Scientific
Danger

OR2212-100G OR2212-25G OR2212-5G
APOSOR2212-100GEA 207 EUR
APOSOR2212-100G APOSOR2212-25G APOSOR2212-5G
2,6-Dibromoaniline 98%
2,6-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250,92 g/mol
Boiling Pt: 262…264 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007638
CAS Number: 608-30-0
EINECS: 210-158-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR