1-Pyrenecarboxaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
OR22205-25G
OR22205-5G
OR22205-100G
APOSOR22205-25GEA
105
EUR
APOSOR22205-25G
APOSOR22205-5G
APOSOR22205-100G
1-Pyrenecarboxaldehyde 98%
1-Pyrenecarboxaldehyde
Formula:
C₁₇H₁₀O Melting Pt: 123…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004139 CAS Number: 3029-19-4 EINECS: 221-196-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...