2-Aminobenzimidazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole
OR22305-100G
OR22305-25G
OR22305-500G
APOSOR22305-100GEA
55.3
EUR
APOSOR22305-100G
APOSOR22305-25G
APOSOR22305-500G
2-Aminobenzimidazole 97%
2-Aminobenzimidazole
Formula:
C₇H₇N₃ MW: 133,15 g/mol Melting Pt: 228…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005596 CAS Number: 934-32-7 EINECS: 213-280-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...