Ethyl (ethoxymethylene)cyanoacetate

Supplier: Apollo Scientific
Danger

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

OR22372-1KG OR22372-250G OR22372-50G
APOSOR22372-1KGEA 0 EUR
APOSOR22372-1KG APOSOR22372-250G APOSOR22372-50G
Ethyl (ethoxymethylene)cyanoacetate
Ethyl (ethoxymethylene)cyanoacetate
Formula: C₈H₁₁NO₃
MW: 169,18 g/mol
Boiling Pt: 190…191 °C/16mmHg
Melting Pt: 47…52 °C
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00009136
CAS Number: 94-05-3
EINECS: 202-299-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR