5-Chloro-2-chloromethyl-1H-benzoimidazole
Supplier: Apollo Scientific
|
Warning
|
OR2243-1G
OR2243-250MG
APOSOR2243-1GEA
119
EUR
APOSOR2243-1G
APOSOR2243-250MG
5-Chloro-2-chloromethyl-1H-benzoimidazole
5-Chloro-2-chloromethyl-1H-benzoimidazole
Formula:
C₈H₆Cl₂N₂ MW: 201,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060503 CAS Number: 20443-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...