2-(2-Quinolyl)malondialdehyde 95+%

Supplier: Apollo Scientific

OR2245-1G OR2245-5G OR2245-25G
APOSOR2245-1GEA 136 EUR
APOSOR2245-1G APOSOR2245-5G APOSOR2245-25G
2-(2-Quinolyl)malondialdehyde 95+%
2-(2-Quinolyl)malondialdehyde
Formula: C₁₂H₉NO₂
MW: 199,21 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD01569245
CAS Number: 40070-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR