2-Brom-4'-pentyloacetophenone 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(Pent-1-yl)phenacyl bromide
OR22471-1G
OR22471-10G
APOSOR22471-1GEA
43.5
EUR
APOSOR22471-1G
APOSOR22471-10G
2-Brom-4'-pentyloacetophenone 95%
2-Brom-4'-pentyloacetophenone
Formula:
C₁₃H₁₇BrO Storage Temperature: Ambient |
CAS Number:
64328-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...