tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride 97%

Supplier: Apollo Scientific

Synonyms: 3-(Aminomethyl)azetidine hydrochloride, 3-BOC protected

OR2249-1G OR2249-5G
APOSOR2249-1GEA 69.6 EUR
APOSOR2249-1G APOSOR2249-5G
tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride 97%
tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride
Formula: C₉H₁₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08752583
CAS Number: 1170108-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR