Chloro(dimethyl)(phenyl)silane 95%

Supplier: Apollo Scientific
Danger

OR22666-100G OR22666-250G
APOSOR22666-100GEA 109 EUR
APOSOR22666-100G APOSOR22666-250G
Chloro(dimethyl)(phenyl)silane 95%
Chloro(dimethyl)(phenyl)silane
Formula: C₈H₁₁ClSi
Boiling Pt: 192…193 °C
Density: 1,017 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000499
CAS Number: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR