4-(Hexyloxy)-3-methoxybenzaldehyde

Supplier: Apollo Scientific

OR2279-10G OR2279-1G OR2279-5G
APOSOR2279-10GEA 469 EUR
APOSOR2279-10G APOSOR2279-1G APOSOR2279-5G
4-(Hexyloxy)-3-methoxybenzaldehyde
4-(Hexyloxy)-3-methoxybenzaldehyde
Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00021080
CAS Number: 61096-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR