1,3-Benzodioxole-4-carbaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Benzo[d][1,3]dioxole-4-carbaldehyde, 2,3-(Methylenedioxy)benzaldehyde, 1,3-Benzodioxole-4-carboxaldehyde
OR23019-25G
APOSOR23019-25GEA
151
EUR
APOSOR23019-25G
1,3-Benzodioxole-4-carbaldehyde
1,3-Benzodioxole-4-carbaldehyde
Formula:
C₈H₆O₃ MW: 150,1300 g/mol Boiling Pt: 80 °C/0,3mmHg Melting Pt: 34…36 °C Density: 1,312 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016902 CAS Number: 7797-83-3 EINECS: 232-252-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...