2-(Bromomethyl)benzothiazole

Supplier: Apollo Scientific
Danger

OR23090-1G OR23090-5G OR23090-50MG
APOSOR23090-1GEA 65.5 EUR
APOSOR23090-1G APOSOR23090-5G APOSOR23090-50MG
2-(Bromomethyl)benzothiazole
2-(Bromomethyl)benzothiazole
Formula: C₈H₆BrNS
MW: 228,11 g/mol
Melting Pt: 47…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00461195
CAS Number: 106086-78-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR