2-Bromobenzothiazole 98%
Supplier: Apollo Scientific
|
Danger
|
OR23092-25G
OR23092-5G
APOSOR23092-25GEA
236
EUR
APOSOR23092-25G
APOSOR23092-5G
2-Bromobenzothiazole 98%
2-Bromobenzothiazole
Formula:
C₇H₄BrNS MW: 214.09 g/mol Melting Pt: 39…43 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD02681887 CAS Number: 2516-40-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...